Assay Details
| Gene Symbol: Cox7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007749.3 | NP_031775.1 | 1 - 2 | 192 | 99 | |
| GenBank mRNA | AK002494.1 | - | 1 - 2 | 159 | 99 | |
| AK027989.1 | - | 1 - 2 | 154 | 99 | ||
| AK131871.1 | - | 1 - 2 | 188 | 99 | ||
| AK153444.1 | - | 1 - 2 | 164 | 99 | ||
| AK168077.1 | - | 1 - 2 | 180 | 99 | ||
| BB696274.1 | - | 1 - 2 | 202 | 99 | ||
| BC010772.1 | - | 1 - 2 | 174 | 4216583 | 99 | |
| BC086792.1 | - | 1 - 2 | 164 | 6411273 | 99 | |
| X52940.1 | - | 1 - 2 | 158 | 99 | ||
| Gene Symbol: Gm12338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110477.1 | 1 | 349 | 99 | ||
| GenBank mRNA | AK137335.1 | - | 99 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196856 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999841 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999838 | NR_028042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999838 | CA533111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999838 | AK137335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999838 | AK002494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999838 | AK027989.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081005 | AV056265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081005 | NM_007749.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53687 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125277 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999841 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999838 | NR_028042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999838 | CA533111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999838 | AK137335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999838 | AK002494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999838 | AK027989.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081005 | AV056265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081005 | NM_007749.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P136153 | AK168077.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P136153 | BC010772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P136153 | X52940.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29196866 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125277 | - | - |
| Whole Mouse Genome G4122A | A_52_P136153 | AK168077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P136153 | BC010772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P136153 | X52940.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10593823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10593797 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10593794 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10593859 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10593812 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29196855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29196782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10593853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10593825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10822365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10993789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10704950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10676562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29196830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10593814 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388462 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10410921 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442922 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388461 | - | - |
| Mouse Genome 430 2.0 Array | 1459884_at | - | - |
| Mouse Genome 430 2.0 Array | 1448112_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459884_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448112_at | - | - |
| Mouse Genome 430 2.0 Array | 1459885_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459885_PM_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300015861.mm.1 | NM_007749.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015861.mm.1 | NM_007749.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015861.mm.1 | TC1300002382.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300015866.mm.1 | NM_007749.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015866.mm.1 | NM_007749.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300015866.mm.1 | TC1300002382.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1459885_PM_s_at | - | - |
| Mouse Expression Set 430 | 1448112_PM_at | - | - |
| Mouse Expression Set 430 | 1459885_s_at | - | - |
| Mouse Expression Set 430 | 1459884_at | - | - |
| Mouse Expression Set 430 | 1459884_PM_at | - | - |
| Mouse Expression Set 430 | 1448112_at | - | - |
| Murine Genome U74v2 Set | 99660_f_at | - | - |
| Murine Genome U74v2 Set | 99661_r_at | - | - |
| Murine Genome U74v2 Set | 99661_PM_r_at | - | - |
| Murine Genome U74v2 Set | 99660_PM_f_at | - | - |
| Murine Genome U74v2 Set | 95896_at | - | - |
| Murine Genome U74v2 Set | 95896_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459884_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448112_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459885_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448112_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459885_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459884_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558196 | - | - |
| ABI Mouse Genome Survey Microarray | 896586 | - | - |
| ABI Mouse Genome Survey Microarray | 896854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2470047 | - | - |
| MouseWG-6 v2 Expression BeadChip | 107050706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107050706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2470047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2647188 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2647188 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259219 | - | - |
| mouse-6 v1.1 (Illumina) | GI_6680990-S | - | - |
| mouse-6 v1.1 (Illumina) | scl012867.3_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107050706 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012867.3_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107050706 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2647188 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2647188 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012867.3_17-S | - | - |

Cox7c
Genomic Map