Assay Details
| Gene Symbol: 2210408I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081353.1 | NP_001074822.1 | 2 - 3 | 229 | 85 | |
| NM_001145676.1 | NP_001139148.1 | 2 - 3 | 229 | 85 | ||
| GenBank mRNA | AK008859.1 | - | 2 - 3 | 215 | 85 | |
| AK050886.1 | - | 2 - 3 | 215 | 85 | ||
| AK143619.1 | - | 2 - 3 | 167 | 85 | ||
| AK165312.1 | - | 2 - 3 | 226 | 85 | ||
| BB649442.1 | - | 2 - 3 | 229 | 85 | ||
| BC089469.1 | - | 2 - 3 | 211 | 85 | ||
| BC151066.1 | - | 2 - 3 | 211 | 9087979 | 85 | |
| BC157949.1 | - | 2 - 3 | 211 | 9088124 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

2210408I21Rik
Genomic Map