Assay Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127177.1 | NP_001120649.1 | 3 - 4 | 375 | 140 | |
| NM_008977.3 | NP_033003.1 | 3 - 4 | 375 | 140 | ||
| GenBank mRNA | AK014528.1 | - | 3 - 4 | 343 | 140 | |
| AK076072.1 | - | 3 - 4 | 328 | 140 | ||
| AK132013.1 | - | 3 - 4 | 338 | 140 | ||
| BC008269.1 | - | 3 - 4 | 349 | 3602983 | 140 | |
| CA560046.1 | - | 3 - 4 | 375 | 140 | ||
| M80739.1 | - | 3 - 4 | 330 | 140 | ||
| M81477.1 | - | 3 - 4 | 330 | 140 | ||
| S52655.1 | - | 3 - 4 | 325 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn2
Genomic Map