Assay Details
| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133254.3 | NP_573517.1 | 8 - 9 | 1017 | 63 | |
| GenBank mRNA | AK128900.1 | - | 8 - 9 | 1031 | 63 | |
| AY033886.1 | - | 8 - 9 | 1027 | 63 | ||
| BC022226.1 | - | 8 - 9 | 1017 | 4235707 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007170 | AK128900.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007170 | BC022226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007170 | NM_133254.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007170 | BC061499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007170 | BC094040.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | AK004109.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | AK053965.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | AK168924.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | BC119523.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | BC127151.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593625 | NM_021453.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47662 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146372 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007170 | AK128900.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007170 | BC022226.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007170 | NM_133254.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007170 | BC061499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007170 | BC094040.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509816 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06363815 | - | - |
| Whole Mouse Genome G4122A | A_52_P407068 | BC022226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407068 | BC017158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407068 | BC024080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407068 | AK154566.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106294 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105556 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146372 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06270088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06270071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06270081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06270091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06504997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06510377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06510402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06060808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06482927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06510413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06060811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06332005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06510403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06504998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06270085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06504999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557979 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419166_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455005_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557975 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568376 | - | - |
| Mouse Genome 430 2.0 Array | 1419166_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419166_at | - | - |
| Mouse Genome 430 2.0 Array | 1455005_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455005_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246787_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 246787_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700016426.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016426.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016426.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016427.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016427.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016427.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016424.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016424.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016424.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016425.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016425.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016425.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016435.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016435.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016435.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016411.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016411.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016411.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016428.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016428.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016428.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016412.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016412.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016412.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016429.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016429.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016429.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016433.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016433.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016433.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016432.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016432.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016432.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016420.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016420.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016420.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016422.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016422.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016422.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016431.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016431.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016431.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016434.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016434.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016434.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016423.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016423.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016423.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016414.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016414.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016414.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016408.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016408.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016408.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016419.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016419.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016419.mm.1 | NM_133254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016430.mm.1 | TC0700001880.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016430.mm.1 | NM_133254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016430.mm.1 | NM_133254.3 | Refseq |
| Mouse Expression Set 430 | 1455005_PM_s_at | - | - |
| Mouse Expression Set 430 | 1419166_at | - | - |
| Mouse Expression Set 430 | 1455005_s_at | - | - |
| Mouse Expression Set 430 | 1419166_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455005_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455005_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419166_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419166_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419166_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419166_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455005_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455005_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771938 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242535 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771938 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242535 | - | - |
| mouse-6 v1.1 (Illumina) | scl32002.10.1_40-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0246787.14_176-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480148 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400242 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0246787.14_176-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32002.10.1_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480148 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400242 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242535 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242535 | - | - |

Slc5a2
Genomic Map