Assay Details
| Gene Symbol: Smpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019476.2 | - | 1 - 2 | 154 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019476.2 transcripts.
Set Membership: |
|

Smpd3
Genomic Map