Assay Details
Gene Symbol: Zfp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011758.2 | NP_035888.2 | 2 - 3 | 279 | 73 | |
GenBank mRNA | AK045944.1 | - | 2 - 3 | 280 | 73 | |
AK141135.1 | - | 2 - 3 | 268 | 73 | ||
BC108271.1 | - | 2 - 3 | 219 | 30102085 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
