Assay Details
| Gene Symbol: Ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027159.2 | NP_081435.1 | 2 - 3 | 550 | 88 | |
| GenBank mRNA | AK010002.1 | - | 2 - 3 | 244 | 88 | |
| AK154179.1 | - | 2 - 3 | 250 | 88 | ||
| AK162237.1 | - | 2 - 3 | 163 | 88 | ||
| BC019430.1 | - | 2 - 3 | 551 | 3491825 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc115
Genomic Map