Assay Details
| Gene Symbol: Lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029609.1 | NP_083885.1 | 5 - 6 | 687 | 75 | |
| GenBank mRNA | AK009207.1 | - | 5 - 6 | 685 | 75 | |
| BC065789.1 | - | 5 - 6 | 663 | 832338 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lhpp
Genomic Map