Assay Details
| Gene Symbol: Krt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010664.2 | NP_034794.2 | 5 - 6 | 995 | 70 | |
| GenBank mRNA | AK145413.1 | - | 5 - 6 | 985 | 70 | |
| AK167072.1 | - | 5 - 6 | 984 | 70 | ||
| AK167265.1 | - | 5 - 6 | 979 | 70 | ||
| AK167432.1 | - | 5 - 6 | 982 | 70 | ||
| AK167469.1 | - | 5 - 6 | 985 | 70 | ||
| AW539037.2 | - | 2 - 3 | 70 | |||
| BC020474.1 | - | 5 - 6 | 985 | 70 | ||
| BC089022.1 | - | 5 - 6 | 957 | 30701486 | 70 | |
| M11686.1 | - | 5 - 6 | 986 | 70 | ||
| M36376.1 | - | 5 - 6 | 981 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt18
Genomic Map