Assay Details
Gene Symbol: Cyp2b23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081148.1 | NP_001074617.1 | 6 - 7 | 965 | 121 | |
GenBank mRNA | BC100376.1 | - | 1 - 2 | 93 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
