Assay Details
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC037681.1 | - | 2 - 3 | 1979 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
