Assay Details
| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198424.3 | NP_940816.1 | 1 - 2 | 403 | 171 | |
| GenBank mRNA | AB271216.1 | - | 1 - 2 | 229 | 171 | |
| AK136955.1 | - | 1 - 2 | 404 | 171 | ||
| AM712358.1 | - | 1 - 2 | 229 | 171 | ||
| BC061259.1 | - | 1 - 2 | 373 | 30251946 | 171 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507196 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507198 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507199 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182690 | BC061259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182690 | AK037138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182690 | AK136955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182690 | NM_198424.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020661 | BC061259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020661 | AK037138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020661 | AK136955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020661 | AK208652.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020661 | NM_198424.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020656 | AK136955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020656 | NM_198424.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437204 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145329 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020656 | AK136955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020656 | NM_198424.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020661 | BC061259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020661 | AK037138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020661 | AK136955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020661 | AK208652.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020661 | NM_198424.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182690 | BC061259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182690 | AK037138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182690 | AK136955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182690 | NM_198424.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25507214 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145329 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01631834 | - | - |
| Whole Mouse Genome G4122A | A_51_P188602 | BC061259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188602 | AK037138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188602 | AK136955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188602 | AK208652.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P188602 | NM_198424.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114322 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06626086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507198 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507196 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25507199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05930189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05930186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06217474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507230 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557755 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434064_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557754 | - | - |
| Mouse Genome 430 2.0 Array | 1434064_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434064_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269999_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269999_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700016011.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016011.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016011.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016015.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016015.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016015.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016017.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016017.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016017.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016016.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016016.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016016.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016018.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016018.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016018.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016012.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016012.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016012.mm.1 | NM_198424.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016014.mm.1 | TC0700001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016014.mm.1 | NM_198424.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016014.mm.1 | NM_198424.3 | Refseq |
| Mouse Expression Set 430 | 1434064_at | - | - |
| Mouse Expression Set 430 | 1434064_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434064_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434064_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 440174 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2030427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1260367 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260367 | - | - |
| mouse-6 v1.1 (Illumina) | scl32018.3_67-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32018.3_67-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1260367 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260367 | - | - |

Orai3
Genomic Map