Assay Details
Gene Symbol: Gbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039647.2 | NP_001034736.2 | 10 - 11 | 1737 | 107 | |
GenBank mRNA | BC111039.1 | - | 10 - 11 | 1734 | 5349539 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042813 | BC111039.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042813 | BI655333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042813 | NM_001039647.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74156 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343539 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334232 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576806 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144762 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273514 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7739 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042813 | BC111039.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042813 | BI655333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042813 | NM_001039647.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23954276 | - | - |
Whole Mouse Genome G4122A | A_51_P327405 | BC080665.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327405 | BC031475.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327405 | NM_029509.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327405 | AK017988.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04905899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04811886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954951 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10532009 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532017 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532023 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532021 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532022 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531998 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532016 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532004 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532018 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532024 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532006 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532001 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10531994 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10532019 | - | - |
Affymetrix Mouse Clariom S Array | PSR0500025636.mm.1 | TC0500002896.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025635.mm.1 | TC0500002896.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025638.mm.1 | TC0500002896.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025633.mm.1 | TC0500002896.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025634.mm.1 | TC0500002896.mm.1 | MTA TC |
Mouse Expression Set 430 | 1421104_at | - | - |
Mouse Expression Set 430 | 1421104_PM_at | - | - |
Mouse Expression Set 430 | 1447927_PM_at | - | - |
Mouse Expression Set 430 | 1447927_at | - | - |
Murine Genome U74v2 Set | 160523_PM_at | - | - |
Murine Genome U74v2 Set | 160523_at | - | - |
Mouse Genome 430A 2.0 Array | 1421104_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421104_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 675464 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242854 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2837493 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837493 | - | - |
