Assay Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008597.3 | NP_032623.1 | 2 - 3 | 171 | 71 | |
GenBank mRNA | AK018719.1 | - | 2 - 3 | 172 | 71 | |
AK159591.1 | - | 2 - 3 | 175 | 71 | ||
BC018338.1 | - | 2 - 3 | 63 | 71 | ||
BC036991.1 | - | 2 - 3 | 167 | 4990627 | 71 | |
BC069922.1 | - | 2 - 3 | 161 | 4984262 | 71 | |
BC079478.1 | - | 2 - 3 | 162 | 4950210 | 71 | |
BY702829.1 | - | 2 - 3 | 172 | 71 | ||
D00613.1 | - | 2 - 3 | 114 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
