Assay Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008597.3 | NP_032623.1 | 3 - 4 | 244 | 62 | |
GenBank mRNA | AK018719.1 | - | 3 - 4 | 245 | 62 | |
AK159591.1 | - | 3 - 4 | 248 | 62 | ||
AK159931.1 | - | 3 - 4 | 245 | 62 | ||
BC018338.1 | - | 3 - 4 | 136 | 62 | ||
BC036991.1 | - | 3 - 4 | 240 | 4990627 | 62 | |
BC069922.1 | - | 3 - 4 | 234 | 4984262 | 62 | |
BC079478.1 | - | 3 - 4 | 235 | 4950210 | 62 | |
BY702829.1 | - | 3 - 4 | 245 | 62 | ||
D00613.1 | - | 3 - 4 | 187 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
