Assay Details
| Gene Symbol: Acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019736.3 | NP_062710.2 | 2 - 3 | 271 | 167 | |
| GenBank mRNA | AB028898.1 | - | 2 - 3 | 175 | 167 | |
| AJ238893.1 | - | 2 - 3 | 226 | 167 | ||
| AK002892.1 | - | 2 - 3 | 235 | 167 | ||
| AK009717.1 | - | 2 - 3 | 234 | 167 | ||
| AK076513.1 | - | 3 - 4 | 325 | 225 | ||
| AK088082.1 | - | 2 - 3 | 234 | 167 | ||
| AK168713.1 | - | 2 - 3 | 273 | 167 | ||
| AK170876.1 | - | 2 - 3 | 234 | 167 | ||
| BC021763.1 | - | 2 - 3 | 261 | 4458575 | 167 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot9
Genomic Map