Assay Details
Gene Symbol: Tpbpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026429.4 | NP_080705.1 | 4 - 5 | 440 | 166 | |
GenBank mRNA | AK005433.1 | - | 4 - 5 | 440 | 166 | |
AK018793.1 | - | 4 - 5 | 439 | 166 | ||
AK018794.1 | - | 4 - 5 | 438 | 166 | ||
AK018795.1 | - | 4 - 5 | 440 | 166 | ||
AK018798.1 | - | 4 - 5 | 440 | 166 | ||
AK018799.1 | - | 4 - 5 | 440 | 166 | ||
AK018804.1 | - | 4 - 5 | 441 | 166 | ||
AK018805.1 | - | 4 - 5 | 440 | 166 | ||
AK018808.1 | - | 4 - 5 | 441 | 166 | ||
AK018809.1 | - | 4 - 5 | 439 | 166 | ||
AK018810.1 | - | 4 - 5 | 440 | 166 | ||
AK018817.1 | - | 4 - 5 | 440 | 166 | ||
AK018820.1 | - | 4 - 5 | 440 | 166 | ||
AK018824.1 | - | 4 - 5 | 440 | 166 | ||
AK018827.1 | - | 4 - 5 | 439 | 166 | ||
AY034576.1 | - | 4 - 5 | 415 | 166 | ||
BC100333.1 | - | 4 - 5 | 425 | 30787126 | 166 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | BC099441.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | BC100333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018805.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018808.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018809.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | BC099441.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | BC100333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018805.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018808.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130808 | AK018809.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533164 | AK142146.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533164 | NM_008700.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533164 | X75415.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718482 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | BC099441.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | BC100333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018805.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018808.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018809.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | BC099441.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | BC100333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018805.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018808.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130808 | AK018809.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29088000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087980 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29087983 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07896462 | - | - |
Whole Mouse Genome G4122A | A_51_P130808 | BC099441.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | BC100333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018805.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018808.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018809.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | BC099441.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | BC100333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018805.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018808.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P130808 | AK018809.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10914013 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10914011 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10914016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29088000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29087942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087949 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11111964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10897170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10914020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087992 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29088091 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10914023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10914011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087971 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29087995 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10409873 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409872 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409871 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416169_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456246_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10409869 | - | - |
Mouse Genome 430 2.0 Array | 1416169_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416169_at | - | - |
Mouse Genome 430 2.0 Array | 1456246_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1456246_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116913_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116913_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300013745.mm.1 | NM_026429.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013745.mm.1 | NM_026429.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013745.mm.1 | TC1300002142.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416169_at | - | - |
Mouse Expression Set 430 | 1456246_PM_x_at | - | - |
Mouse Expression Set 430 | 1456246_x_at | - | - |
Mouse Expression Set 430 | 1416169_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416169_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1456246_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1456246_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1416169_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456246_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456246_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416169_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416169_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1740072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001740072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250714 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250714 | - | - |
mouse-6 v1.1 (Illumina) | scl43863.4.1_3-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1740072 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl43863.4.1_3-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2867252 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250714 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2867252 | - | - |
