Assay Details
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016897.3 | NP_058593.2 | 3 - 4 | 406 | 108 | |
GenBank mRNA | AB021122.1 | - | 3 - 4 | 386 | 108 | |
AK013976.1 | - | 3 - 4 | 352 | 108 | ||
AK146516.1 | - | 3 - 4 | 372 | 108 | ||
BC037022.1 | - | 3 - 4 | 383 | 5366968 | 108 | |
BC062963.1 | - | 3 - 4 | 342 | 5692352 | 108 | |
BC069903.1 | - | 3 - 4 | 376 | 5290493 | 108 | |
BC096669.1 | - | 3 - 4 | 375 | 5056242 | 108 | |
BC132019.1 | - | 3 - 4 | 355 | 40130296 | 108 | |
BC132319.1 | - | 3 - 4 | 355 | 40130596 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
