Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00004691.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004691.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004691.mm.1 |
TC0X00000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004686.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004686.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004686.mm.1 |
TC0X00000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004694.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004694.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004694.mm.1 |
TC0X00000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004696.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004696.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004696.mm.1 |
TC0X00000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004695.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004695.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004695.mm.1 |
TC0X00000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004674.mm.1 |
NM_052835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004674.mm.1 |
NM_052835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004674.mm.1 |
TC0X00000704.mm.1 |
MTA TC |