| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600012839.mm.1 |
NM_001082546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012839.mm.1 |
NM_001082546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012839.mm.1 |
TC1600001573.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012836.mm.1 |
NM_001082546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012836.mm.1 |
NM_001082546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012836.mm.1 |
TC1600001573.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012844.mm.1 |
TC1600001573.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012840.mm.1 |
NM_001082546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012840.mm.1 |
NM_001082546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012840.mm.1 |
TC1600001573.mm.1 |
MTA TC |