Assay Details
| Gene Symbol: Snrpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC051470.1 | - | 2 - 3 | 149 | 1244608 | 150 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051470.1 transcripts.
Set Membership: |
|

Snrpg
Genomic Map