Assay Details
Gene Symbol: Cox5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009942.2 | NP_034072.2 | 2 - 3 | 277 | 70 | |
GenBank mRNA | AK008339.1 | - | 2 - 3 | 198 | 70 | |
AK131673.1 | - | 2 - 3 | 198 | 70 | ||
AK135174.1 | - | 2 - 3 | 206 | 70 | ||
BC005564.1 | - | 217 | 70 | |||
BC096048.1 | - | 2 - 3 | 192 | 3465479 | 70 | |
BC099474.1 | - | 2 - 3 | 206 | 30933585 | 70 | |
BU757937.1 | - | 2 - 3 | 70 | |||
CJ075429.1 | - | 2 - 3 | 282 | 70 | ||
X53157.1 | - | 2 - 3 | 186 | 70 |
Gene Symbol: Gm11273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC023228.1 | - | 205 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
