Assay Details
| Gene Symbol: Eif4ebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007918.3 | NP_031944.3 | 1 | 86 | 147 | |
| GenBank mRNA | BC002045.1 | - | 1 | 54 | 3592595 | 147 |
| BY318447.1 | - | 1 | 87 | 147 | ||
| U28656.1 | - | 1 | 54 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC002045.1, NM_007918.3, U28656.1 transcripts.
Set Membership: |
|

Eif4ebp1
Genomic Map