Assay Details
| Gene Symbol: Ube2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009458.4 | NP_033484.3 | 6 | 138 | ||
| GenBank mRNA | AK147785.1 | - | 6 | 138 | ||
| BE944184.1 | - | 1 | 384 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube2b
Genomic Map