Assay Details
Gene Symbol: Sprr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011470.2 | NP_035600.1 | 2 | 556 | 122 | |
GenBank mRNA | AJ005562.1 | - | 2 | 539 | 122 | |
AK008778.1 | - | 2 | 557 | 122 | ||
AY158988.1 | - | 2 | 557 | 122 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AK008778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AY158988.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AK008778.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | AY158988.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902465 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156415 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AK008778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AY158988.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AK008778.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | AY158988.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330963 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22330959 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156415 | - | - |
Whole Mouse Genome G4122A | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | AK008778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | AY158988.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | AJ005562.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | AK008778.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | AY158988.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P435588 | NM_011470.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139156 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107082 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156415 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03033325 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02399039 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02399031 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02399035 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02399038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22330999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03006716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03033324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02778614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03169747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03087225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03087221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03087230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03087223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22330952 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10493866 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10493865 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420771_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10493864 | - | - |
Mouse Genome 430 2.0 Array | 1420771_at | - | - |
Mouse Genome 430 2.0 Array | 1420771_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20758_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20758_PM_at | - | - |
Mouse Expression Set 430 | 1420771_at | - | - |
Mouse Expression Set 430 | 1420771_PM_at | - | - |
Murine Genome U74v2 Set | 101761_PM_f_at | - | - |
Murine Genome U74v2 Set | 101761_f_at | - | - |
Murine Genome U74v2 Set | 95793_at | - | - |
Murine Genome U74v2 Set | 95793_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420771_at | - | - |
Mouse Genome 430A 2.0 Array | 1420771_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420771_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420771_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 375832 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1340458 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001340458 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626516 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2626516 | - | - |
mouse-6 v1.1 (Illumina) | scl020758.2_247-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1340458 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl020758.2_247-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340458 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2626516 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626516 | - | - |
