Assay Details
| Gene Symbol: Cox8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007750.2 | NP_031776.1 | 1 | 119 | 143 | |
| GenBank mRNA | AK002218.1 | - | 1 | 79 | 143 | |
| AK008594.1 | - | 1 | 70 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002218.1,AK008594.1,NM_007750.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cox8a
Genomic Map