Assay Details
| Gene Symbol: Cyp4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010011.3 | NP_034141.3 | 1 | 112 | 169 | |
| GenBank mRNA | AB017785.1 | - | 1 | 102 | 169 | |
| AB018421.1 | - | 1 | 106 | 169 | ||
| AK002528.1 | - | 1 | 113 | 169 | ||
| AK098101.1 | - | 1 | 110 | 169 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB017785.1, AB018421.1, AK002528.1, NM_010011.3 transcripts.
Set Membership: |
|

Cyp4a10
Genomic Map