Assay Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282032.1 | NP_001268961.1 | 1 | 78 | 117 | |
NM_011826.4 | NP_035956.1 | 1 | 78 | 117 | ||
GenBank mRNA | AF023482.1 | - | 1 | 50 | 117 | |
AF023483.1 | - | 1 | 50 | 117 | ||
AK088746.1 | - | 1 | 60 | 117 | ||
AK135138.1 | - | 1 | 40 | 117 | ||
BY253818.1 | - | 1 | 67 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF023482.1, AK088746.1, AK135138.1, NM_001282032.1, NM_011826.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
positive regulation of actin cytoskeleton reorganization
protein binding
protein domain specific binding
interleukin-1 binding
protein N-terminus binding
cell surface receptor signaling pathway
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
positive regulation of actin cytoskeleton reorganization
protein binding
protein domain specific binding
interleukin-1 binding
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116889 | BC006688.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116889 | BC098225.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116889 | BC147481.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116889 | AK002256.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116889 | AK010633.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096187 | BC006688.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096187 | BC098225.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096187 | AK135138.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096187 | AK002256.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617092 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171424 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096187 | BC006688.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096187 | BC098225.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096187 | AK135138.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096187 | AK002256.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116889 | BC006688.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116889 | BC098225.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116889 | BC147481.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116889 | AK002256.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116889 | AK010633.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437697 | BC006688.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437697 | BC098225.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437697 | AK135138.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437697 | AF023483.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437697 | AK002256.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322823 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322820 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22322855 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171424 | - | - |
Whole Mouse Genome G4122A | A_52_P466741 | BC006688.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466741 | BC098225.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466741 | BC147481.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466741 | AK135138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466741 | AF023483.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P437697 | BC006688.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P437697 | BC098225.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P437697 | AK135138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P437697 | AF023483.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P437697 | AK002256.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P572818 | AF023483.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139331 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136417 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171424 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02712876 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02524216 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02524222 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02898598 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02524218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322819 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02524221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02821994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02898598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02701575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02524217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322829 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02524214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322816 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22322809 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02524216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10499710 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499715 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499712 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499706 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499711 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499708 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499709 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499707 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478942 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478940 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499714 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499713 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426187_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438180_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10499705 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10478938 | - | - |
Mouse Genome 430 2.0 Array | 1438180_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1438180_x_at | - | - |
Mouse Genome 430 2.0 Array | 1426187_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1426187_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300018274.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018274.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018268.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018268.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018268.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018275.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018275.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018275.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018279.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018279.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018279.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018267.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018267.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018267.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018271.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018271.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018271.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018269.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018269.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018269.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018270.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018270.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018270.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018272.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018272.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018272.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018273.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018273.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018276.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018276.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018276.mm.1 | TC0300002360.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300018280.mm.1 | NM_001282032.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018280.mm.1 | NM_011826.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300018280.mm.1 | TC0300002360.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426187_PM_a_at | - | - |
Mouse Expression Set 430 | 1426187_a_at | - | - |
Mouse Expression Set 430 | 1438180_PM_x_at | - | - |
Mouse Expression Set 430 | 1438180_x_at | - | - |
Murine Genome U74v2 Set | 101096_PM_s_at | - | - |
Murine Genome U74v2 Set | 101096_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1438180_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1426187_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1438180_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1426187_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438180_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426187_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438180_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426187_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 860392 | - | - |
ABI Mouse Genome Survey Microarray | 920381 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4150670 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4150670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004150670 | - | - |
mouse-6 v1.1 (Illumina) | scl023897.2_20-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4150670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl023897.2_20-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2972392 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2750013 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2972392 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl023897.2_20-S | - | - |
