Assay Details
| Gene Symbol: Acnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164565.1 | NP_001158037.1 | 3 | 933 | 158 | |
| GenBank mRNA | DQ469311.1 | - | 3 | 933 | 158 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
thiolester hydrolase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
thiolester hydrolase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115836 | NM_001164565.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202264 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115836 | NM_001164565.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03410695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03584153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03940089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03940093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04077907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03737413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03508835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03648024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22905273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04077905 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512893 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512892 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400021451.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021451.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021451.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021453.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021453.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021453.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021452.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021452.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021452.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021445.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021445.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021445.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021447.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021447.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021447.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021449.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021449.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021449.mm.1 | NM_001164565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021450.mm.1 | TC0400002634.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021450.mm.1 | NM_001164565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021450.mm.1 | NM_001164565.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 349503 | - | - |
| ABI Mouse Genome Survey Microarray | 867311 | - | - |

Acnat1
Genomic Map