Assay Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207301.2 | NP_997184.1 | 5 | 2278 | 138 | |
| GenBank mRNA | AK017448.1 | - | 1 | 1420 | 138 | |
| BC031769.1 | - | 5 | 2271 | 4235618 | 138 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991290 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991295 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076159 | BC031769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076159 | AK172079.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076159 | AK179336.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076159 | NM_207301.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562706 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132464 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149772 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171316 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076159 | BC031769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076159 | AK172079.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076159 | AK179336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076159 | NM_207301.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30991299 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149772 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694830 | - | - |
| Whole Mouse Genome G4122A | A_52_P418602 | BC031769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418602 | AK017448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418602 | NM_207301.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321903 | AK084688.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110274 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149772 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12731974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12782376 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12731972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991290 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30991289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30991265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12649785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12677520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12609363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12782377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12507317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12942613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13013387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13049765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12905672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12703761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12609365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12815858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30991336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437177 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430312_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460446_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437174 | - | - |
| Mouse Genome 430 2.0 Array | 1430312_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460446_at | - | - |
| Mouse Genome 430 2.0 Array | 1460446_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430312_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71446_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71446_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460446_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430312_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430312_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460446_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101770021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 870148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000870148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2473118 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220592 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2473118 | - | - |
| mouse-6 v1.1 (Illumina) | scl48836.8_442-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101770021 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 870148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48836.8_442-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101770021 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2473122 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220592 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2473122 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl48836.8_442-S | - | - |

Wrb
Genomic Map