Assay Details
Gene Symbol: Zfp85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001130.2 | NP_001001130.1 | 4 | 1483 | 143 | |
GenBank mRNA | BC055337.1 | - | 4 | 1483 | 6413269 | 143 |
BC055756.1 | - | 4 | 1483 | 6413269 | 143 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967301 | NM_001001130.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967301 | BC055337.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967301 | BC055756.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11504 | NM_001001130.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11504 | BC055337.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11504 | BC055756.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967301 | NM_001001130.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967301 | BC055337.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967301 | BC055756.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11504 | NM_001001130.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11504 | BC055337.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11504 | BC055756.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109715 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109718 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29109734 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10601731 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10597362 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10601718 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10601720 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11003456 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10597359 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11003458 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10601725 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10597364 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10597366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29109651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10468537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109673 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10601725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10757149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11003456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11089163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11003464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109693 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10829018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10916019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29109738 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10410378 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410377 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10410376 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10410375 | - | - |
Mouse Genome 430 2.0 Array | 1455503_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455503_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300014768.mm.1 | NM_001001130.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014768.mm.1 | NM_001001130.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014768.mm.1 | TC1300002260.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300014769.mm.1 | NM_001001130.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014769.mm.1 | NM_001001130.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014769.mm.1 | TC1300002260.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300014767.mm.1 | NM_001001130.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014767.mm.1 | NM_001001130.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014767.mm.1 | TC1300002260.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300014766.mm.1 | NM_001001130.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014766.mm.1 | NM_001001130.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014766.mm.1 | TC1300002260.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455503_at | - | - |
Mouse Expression Set 430 | 1455503_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 767073 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161206 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3161206 | - | - |
mouse-6 v1.1 (Illumina) | GI_47564116-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3161206 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161206 | - | - |
