Assay Details
| Gene Symbol: Rpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014593.1 | - | 2 | 2618 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331862 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010341 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139673 | AK153486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139673 | AK169053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139673 | BC025077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139673 | BC069896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139673 | AK008422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125075 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967076 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139673 | AK153486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139673 | AK169053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139673 | BC025077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139673 | BC069896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139673 | AK008422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010341 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394524 | AK153486.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394524 | AK164445.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394524 | AK169053.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394524 | BC021402.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394524 | BC025077.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23331845 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125075 | - | - |
| Whole Mouse Genome G4122A | A_52_P566208 | AK014593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394524 | AK153486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394524 | AK164445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394524 | AK169053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394524 | BC021402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394524 | BC025077.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129725 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124787 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128063 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133993 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04062112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04062117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04062116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03450851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04062108 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04062109 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23331868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23331802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03467539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03716293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03263167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04015716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03350636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03448751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03809042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03961123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03809041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03452891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23331825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10594143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10502746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10502747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517460 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417615_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10594142 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451301 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517457 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528544 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10502745 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454097 | - | - |
| Mouse Genome 430 2.0 Array | 1417615_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417615_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448773_at | - | - |
| Mouse Genome 430 2.0 Array | 1448773_PM_at | - | - |
| Mouse Expression Set 430 | 1448773_PM_at | - | - |
| Mouse Expression Set 430 | 1417615_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448773_at | - | - |
| Mouse Expression Set 430 | 1417615_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448773_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417615_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417615_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448773_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417615_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417615_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 643745 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665229 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665229 | - | - |
| mouse-6 v1.1 (Illumina) | scl067025.2_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067025.2_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780435 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665229 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067025.2_6-S | - | - |

Rpl11
Genomic Map