Assay Details
| Gene Symbol: Apol7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175391.4 | NP_780600.1 | 5 | 992 | 156 | |
| GenBank mRNA | BC038135.1 | - | 1 | 280 | 156 | |
| BC139148.1 | - | 5 | 951 | 8862170 | 156 | |
| BC139149.1 | - | 5 | 951 | 8862171 | 156 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148062 | BC139149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148062 | BC139148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148062 | NM_175391.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148062 | AK075855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148062 | BC038135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148062 | BC139149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148062 | BC139148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148062 | NM_175391.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148062 | AK075855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148062 | BC038135.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30354582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03075645 | - | - |
| Whole Mouse Genome G4122A | A_51_P239177 | NR_033577.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239177 | BC052510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239177 | NM_175391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239177 | AK075855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P239177 | BC038135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407171 | BC139149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407171 | BC139148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407171 | NM_175391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407171 | AK075855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407171 | BC038135.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103899 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133438 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03075645 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12279639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12279616 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12279611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12279611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12279595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12279620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12279616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354583 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430189 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430179 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425040 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430186 | - | - |
| Mouse Genome 430 2.0 Array | 1434152_at | - | - |
| Mouse Genome 430 2.0 Array | 1434152_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014066.mm.1 | NM_175391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014066.mm.1 | NM_175391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014066.mm.1 | TC1500001812.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429272_PM_a_at | - | - |
| Mouse Expression Set 430 | 1434152_PM_at | - | - |
| Mouse Expression Set 430 | 1434152_at | - | - |
| Mouse Expression Set 430 | 1429272_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 372851 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3360215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614563 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2614563 | - | - |
| mouse-6 v1.1 (Illumina) | scl0108956.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0108956.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2614563 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2614563 | - | - |

Apol7c
Genomic Map