Assay Details
| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024173.2 | NP_077135.1 | 1 | 105 | 100 | |
| GenBank mRNA | AK007596.1 | - | 1 | 48 | 100 | |
| AK010131.1 | - | 1 | 43 | 100 | ||
| AK081611.1 | - | 1 | 65 | 100 | ||
| AK168972.1 | - | 1 | 43 | 100 | ||
| CN838691.1 | - | 1 | 108 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007596.1, AK010131.1, AK081611.1, AK168972.1, NM_024173.2 transcripts.
Set Membership: |
|

Atp6v1g1
Genomic Map