Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 4 - 5 | 532 | 94 | |
| GenBank mRNA | AK013295.1 | - | 4 - 5 | 523 | 94 | |
| AK131964.1 | - | 4 - 5 | 553 | 94 | ||
| AK136951.1 | - | 4 - 5 | 556 | 94 | ||
| AK161347.1 | - | 4 - 5 | 499 | 94 | ||
| AK161376.1 | - | 4 - 5 | 553 | 94 | ||
| AK162409.1 | - | 4 - 5 | 553 | 94 | ||
| AK167991.1 | - | 4 - 5 | 553 | 94 | ||
| AK169850.1 | - | 4 - 5 | 567 | 94 | ||
| BC021755.1 | - | 4 - 5 | 537 | 94 | ||
| BC052027.1 | - | 4 - 5 | 559 | 5718637 | 94 | |
| D63784.1 | - | 4 - 5 | 531 | 94 | ||
| U53208.1 | - | 4 - 5 | 487 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnajc2
Genomic Map