Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190334.1 | NP_001177263.1 | 1 - 2 | 49 | 63 | |
| NM_001190336.1 | NP_001177265.1 | 1 - 2 | 49 | 63 | ||
| NM_009835.4 | NP_033965.1 | 2 - 3 | 177 | 63 | ||
| GenBank mRNA | AK089712.1 | - | 2 - 3 | 178 | 63 | |
| BC100756.1 | - | 1 - 2 | 45 | 40046797 | 63 | |
| BC100758.1 | - | 1 - 2 | 45 | 40046794 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089712.1, BC100756.1, BC100758.1, NM_001190334.1, NM_001190336.1, NM_009835.4 transcripts.
Set Membership: |
|

Ccr6
Genomic Map