Assay Details
| Gene Symbol: Mos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020021.2 | NP_064405.2 | 1 | 988 | 92 | |
| GenBank mRNA | AK133089.1 | - | 1 | 988 | 92 | |
| BC137690.1 | - | 1 | 901 | 8860709 | 92 | |
| BC145745.1 | - | 1 | 901 | 40131048 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22694765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22694768 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101078 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22694753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101078 | - | - |
| Whole Mouse Genome G4122A | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | AK133089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | BC137690.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | BC145745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P42332 | NM_020021.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133629 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729958 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03729968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22694765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22694768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22694722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03374043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03865187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04019833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03865185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03865184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03865186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22694703 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10511331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511332 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450775_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511330 | - | - |
| Mouse Genome 430 2.0 Array | 1450775_at | - | - |
| Mouse Genome 430 2.0 Array | 1450775_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17451_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17451_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400017934.mm.1 | NM_020021.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017934.mm.1 | NM_020021.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017934.mm.1 | TC0400002131.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017935.mm.1 | NM_020021.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017935.mm.1 | NM_020021.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017935.mm.1 | TC0400002131.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017936.mm.1 | NM_020021.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017936.mm.1 | NM_020021.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017936.mm.1 | TC0400002131.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450775_PM_at | - | - |
| Mouse Expression Set 430 | 1450775_at | - | - |
| Murine Genome U74v2 Set | 96521_PM_at | - | - |
| Murine Genome U74v2 Set | 161867_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161867_f_at | - | - |
| Murine Genome U74v2 Set | 96521_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450775_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450775_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450775_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450775_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5290619 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290619 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290619 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678538 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2678538 | - | - |
| mouse-6 v1.1 (Illumina) | scl2560.1.1_255-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290619 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl2560.1.1_255-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290619 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2982636 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2982636 | - | - |

Mos
Genomic Map