Assay Details
| Gene Symbol: Mrpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027098.2 | NP_081374.1 | 3 - 4 | 186 | 117 | |
| GenBank mRNA | AK009074.1 | - | 3 - 4 | 187 | 117 | |
| AK089015.1 | - | 3 - 4 | 187 | 117 | ||
| BC004614.1 | - | 2 - 3 | 165 | 3585410 | 117 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK009074.1,AK089015.1,BC004614.1,NM_027098.2 transcripts.
This assay is in the 5'UTR region of the AK009074.1,AK089015.1,BC004614.1,NM_027098.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156460 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103944 | NM_027098.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103944 | AK009074.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103944 | AK089015.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103944 | BC004614.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227088 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113435 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008755 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103944 | NM_027098.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103944 | AK009074.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103944 | AK089015.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103944 | BC004614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P440904 | AK089015.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156463 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113435 | - | - |
| Whole Mouse Genome G4122A | A_51_P440904 | AK089015.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120174 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002555 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00578510 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002556 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20156460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00669491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00766553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00845444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00669492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00766554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00822244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00669497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00669495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00310424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00822247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00845445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20156497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002555 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10345623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345624 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423857_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345620 | - | - |
| Mouse Genome 430 2.0 Array | 1423857_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423857_at | - | - |
| Mouse Genome 430 2.0 Array | 1447651_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447651_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107734_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107734_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100001920.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001920.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001920.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001921.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001921.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001921.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001917.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001917.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001917.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001919.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001919.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001919.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001925.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001925.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001925.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001924.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001924.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001924.mm.1 | TC0100000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001923.mm.1 | NM_027098.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001923.mm.1 | NM_027098.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001923.mm.1 | TC0100000268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423857_at | - | - |
| Mouse Expression Set 430 | 1423857_PM_at | - | - |
| Mouse Expression Set 430 | 1447651_PM_x_at | - | - |
| Mouse Expression Set 430 | 1447651_x_at | - | - |
| Murine Genome U74v2 Set | 96082_at | - | - |
| Murine Genome U74v2 Set | 96082_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423857_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423857_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423857_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423857_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 522734 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1260262 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260262 | - | - |
| mouse-6 v1.1 (Illumina) | scl0107734.5_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700170 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0107734.5_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1260262 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260262 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0107734.5_7-S | - | - |

Mrpl30
Genomic Map