Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600025407.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025407.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025407.mm.1 |
NM_133203.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025409.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025409.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025409.mm.1 |
NM_133203.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025411.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025411.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025411.mm.1 |
NM_133203.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025408.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025408.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025408.mm.1 |
NM_133203.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025412.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025412.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025412.mm.1 |
NM_133203.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025405.mm.1 |
TC0600003250.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025405.mm.1 |
NM_133203.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025405.mm.1 |
NM_133203.5 |
Refseq |