Assay Details
| Gene Symbol: Carns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC023699.1 | - | 1 - 2 | 140 | 5345551 | 79 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023699.1 transcripts.
Set Membership: |
|

Carns1
Genomic Map