Assay Details
| Gene Symbol: Anapc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181394.3 | NP_852059.1 | 2 - 3 | 182 | 89 | |
| GenBank mRNA | AK007332.1 | - | 1 - 2 | 161 | 89 | |
| BC028526.1 | - | 1 - 2 | 147 | 3371464 | 89 | |
| BY347589.1 | - | 2 - 3 | 182 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781232 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781224 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92148 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107305 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26781235 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107305 | - | - |
| Whole Mouse Genome G4122A | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368903 | NM_181394.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368903 | BC028526.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129489 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107305 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781232 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26781226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26781276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07789341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08043074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07500493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07884230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08114080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07521793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07521787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08028856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07500494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08114084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08161273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07884232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08043073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08043070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26781345 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588225 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452586_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588223 | - | - |
| Mouse Genome 430 2.0 Array | 1452586_at | - | - |
| Mouse Genome 430 2.0 Array | 1452586_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69010_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69010_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900009989.mm.1 | NM_181394.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009989.mm.1 | NM_181394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009989.mm.1 | TC0900001290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900009992.mm.1 | NM_181394.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009992.mm.1 | NM_181394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009992.mm.1 | TC0900001290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900009991.mm.1 | NM_181394.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009991.mm.1 | NM_181394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900009991.mm.1 | TC0900001290.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452586_PM_at | - | - |
| Mouse Expression Set 430 | 1452586_at | - | - |
| Murine Genome U74v2 Set | 162466_PM_at | - | - |
| Murine Genome U74v2 Set | 93539_PM_at | - | - |
| Murine Genome U74v2 Set | 162466_at | - | - |
| Murine Genome U74v2 Set | 93539_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452586_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452586_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452586_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452586_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 583790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3800446 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800446 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800446 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220454 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220454 | - | - |
| mouse-6 v1.1 (Illumina) | scl069010.2_129-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800446 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl069010.2_129-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800446 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2796089 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2796089 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl069010.2_129-S | - | - |

Anapc13
Genomic Map