Assay Details
Gene Symbol: Smim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183259.3 | NP_899082.1 | 2 - 3 | 350 | 74 | |
GenBank mRNA | AK004370.1 | - | 2 - 3 | 283 | 74 | |
AK020450.1 | - | 2 - 3 | 312 | 74 | ||
BC032884.1 | - | 2 - 3 | 74 | |||
BC065414.1 | - | 3 - 4 | 338 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | BC065414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AK004370.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AK020450.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AW124215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | BC065414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AK004370.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AK020450.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295104 | AW124215.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117009 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136821 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120628 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | BC065414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AK004370.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AK020450.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AW124215.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | BC065414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AK004370.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AK020450.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295104 | AW124215.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156083 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28156103 | - | - |
Whole Mouse Genome G4122A | A_52_P295104 | BC065414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AK004370.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AK020450.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AW124215.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | BC065414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | NM_183259.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AK004370.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AK020450.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P295104 | AW124215.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113721 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09669070 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09170930 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09398674 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09398665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09398705 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10382695 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382694 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382697 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382696 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10382693 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435378_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10382692 | - | - |
Mouse Genome 430 2.0 Array | 1435378_at | - | - |
Mouse Genome 430 2.0 Array | 1435378_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66528_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66528_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100018142.mm.1 | NM_183259.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018142.mm.1 | NM_183259.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018142.mm.1 | TC1100001911.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100018144.mm.1 | NM_183259.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018144.mm.1 | NM_183259.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018144.mm.1 | TC1100001911.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100018143.mm.1 | NM_183259.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018143.mm.1 | NM_183259.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100018143.mm.1 | TC1100001911.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435378_PM_at | - | - |
Mouse Expression Set 430 | 1435378_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435378_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435378_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 461120 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4590273 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4590273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004590273 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2643326 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2643326 | - | - |
mouse-6 v1.1 (Illumina) | scl40718.3_48-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4590273 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl40718.3_48-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590273 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2643326 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2643326 | - | - |
