Assay Details
| Gene Symbol: Qrfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183424.3 | NP_906269.1 | 1 - 2 | 113 | 74 | |
| GenBank mRNA | AK029144.1 | - | 1 - 2 | 114 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029144.1, NM_183424.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021861 | AK029144.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021861 | NM_183424.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69107 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169737 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021861 | AK029144.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021861 | NM_183424.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21111388 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169737 | - | - |
| Whole Mouse Genome G4122A | A_52_P184414 | AK029144.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P184414 | NM_183424.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122449 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02197951 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02197960 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02197955 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02197953 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02197952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21111360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02099913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02197953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01480311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02197961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01480354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01480348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02197960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01480306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01307538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21111363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10481564 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481562 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481560 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200027927.mm.1 | NM_183424.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027927.mm.1 | TC0200003291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200027929.mm.1 | NM_183424.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027929.mm.1 | TC0200003291.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200027926.mm.1 | NM_183424.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027926.mm.1 | TC0200003291.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 461194 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3610435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222007 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222007 | - | - |
| mouse-6 v1.1 (Illumina) | scl19456.2_692-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19456.2_692-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2824999 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2824999 | - | - |

Qrfp
Genomic Map