| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600018068.mm.1 |
TC0600002312.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600018068.mm.1 |
NR_033635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600018068.mm.1 |
NR_033635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600018067.mm.1 |
TC0600002312.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600018067.mm.1 |
NR_033635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600018067.mm.1 |
NR_033635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600018065.mm.1 |
TC0600002312.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600018065.mm.1 |
NR_033635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600018065.mm.1 |
NR_033635.1 |
Refseq |