Assay Details
| Gene Symbol: Eid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198425.2 | NP_940817.1 | 1 | 729 | 69 | |
| GenBank mRNA | AY251273.1 | - | 1 | 655 | 69 | |
| BC094513.1 | - | 1 | 729 | 2648407 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eid2
Genomic Map