Assay Details
| Gene Symbol: Mex3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198615.2 | NP_941017.2 | 1 - 2 | 985 | 157 | |
| GenBank mRNA | AK149801.1 | - | 1 - 2 | 513 | 157 | |
| BC059858.1 | - | 2 - 3 | 105 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282840 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282869 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093439 | BX514891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093439 | NM_198615.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058737 | BC059858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058737 | BX514891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058737 | NM_198615.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112758 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093439 | BX514891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093439 | NM_198615.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058737 | BC059858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058737 | BX514891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058737 | NM_198615.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07122401 | - | - |
| Whole Mouse Genome G4122A | A_52_P568165 | BC059858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P568165 | NM_198615.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08782501 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08782483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08782494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08831056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08274046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08782501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08629817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08559113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08828367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08246101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08629816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08615693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08458031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08923667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27282897 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370820 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437627_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370818 | - | - |
| Mouse Genome 430 2.0 Array | 1437627_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437627_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237400_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237400_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000018480.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018480.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018480.mm.1 | TC1000002498.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018479.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018479.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018479.mm.1 | TC1000002498.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018477.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018477.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018477.mm.1 | TC1000002498.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018478.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018478.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018478.mm.1 | TC1000002498.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018476.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018476.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018476.mm.1 | TC1000002498.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018475.mm.1 | NM_198615.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018475.mm.1 | NM_198615.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018475.mm.1 | TC1000002498.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437627_at | - | - |
| Mouse Expression Set 430 | 1437627_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437627_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437627_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6860577 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6860577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724699 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724699 | - | - |
| mouse-6 v1.1 (Illumina) | scl0237400.1_111-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0237400.1_111-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724702 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724702 | - | - |

Mex3d
Genomic Map