Assay Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC057593.1 | - | 4 - 5 | 419 | 65 | |
| CA320661.1 | - | 4 - 5 | 420 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp952| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC057593.1 | - | 4 - 5 | 419 | 65 | |
| CA320661.1 | - | 4 - 5 | 420 | 65 | ||
Set Membership: |
|