Assay Details
Gene Symbol: Tas2r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199154.1 | NP_954605.1 | 1 | 878 | 103 | |
GenBank mRNA | BC141027.1 | - | 1 | 943 | 9055639 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000042 | NM_199154.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174362 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000042 | NM_199154.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24851592 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174362 | - | - |
Whole Mouse Genome G4122A | A_52_P213731 | NM_199154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213731 | AF412301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213731 | BC141027.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127018 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174362 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510546 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510537 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510552 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510561 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510539 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510536 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510538 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05510549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24851615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05510558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05510537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05510549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05783140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05836263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24851582 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10548607 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548606 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600025745.mm.1 | TC0600003272.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025745.mm.1 | NM_199154.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025744.mm.1 | TC0600003272.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025746.mm.1 | TC0600003272.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2350397 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2350397 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002350397 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2658741 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2658741 | - | - |
mouse-6 v1.1 (Illumina) | scl0387342.1_267-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2350397 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387342.1_267-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350397 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2658741 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2658741 | - | - |
