Assay Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203320.3 | NP_976065.1 | 2 - 3 | 273 | 98 | |
| GenBank mRNA | AK150314.1 | - | 2 - 3 | 272 | 98 | |
| AK154334.1 | - | 2 - 3 | 272 | 98 | ||
| AY311403.1 | - | 2 - 3 | 271 | 98 | ||
| BC117014.1 | - | 2 - 3 | 228 | 40126333 | 98 | |
| BC117016.1 | - | 2 - 3 | 228 | 40126335 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cxcl3
Genomic Map