Assay Details
| Gene Symbol: Mrgprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203490.3 | NP_987075.1 | 2 | 1281 | 64 | |
| GenBank mRNA | AY762266.1 | - | 2 | 1281 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986873 | NM_203490.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986873 | AY762266.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103465 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177141 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137850 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986873 | NM_203490.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986873 | AY762266.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25612541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01662803 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177141 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137850 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117498 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137850 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177141 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06507836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06507840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06134198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06507847 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06507843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25612615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06139578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06210089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06194288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06194286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06600580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06571307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06194289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06210105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06551137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06139575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25612585 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559391 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559389 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019087.mm.1 | TC0700002118.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019087.mm.1 | NM_203490.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019087.mm.1 | NM_203490.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4070044 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689781 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689781 | - | - |
| mouse-6 v1.1 (Illumina) | scl0211578.1_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070044 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0211578.1_182-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2689781 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689781 | - | - |

Mrgprd
Genomic Map